Skip to Content
Merck
All Photos(1)

Key Documents

124141

Sigma-Aldrich

4-Bromostyrene

contains 0.05% 3,5-di-tert-butylcatechol as inhibitor, 97%

Synonym(s):

1-(4-Bromophenyl)ethylene, 1-Bromo-4-ethenylbenzene, 1-Bromo-4-vinylbenzene, 4-Vinyl-1-bromobenzene, p -Bromostyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4Br
CAS Number:
Molecular Weight:
183.05
Beilstein:
1634204
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

contains

0.05% 3,5-di-tert-butylcatechol as inhibitor

refractive index

n20/D 1.594 (lit.)

bp

89 °C/16 mmHg (lit.)

density

1.4 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Brc1ccc(C=C)cc1

InChI

1S/C8H7Br/c1-2-7-3-5-8(9)6-4-7/h2-6H,1H2

InChI key

WGGLDBIZIQMEGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromostyrene is a para-halogenated styrene derivative. The proton spectra of 4-bromostyrene exhibits dipolar couplings consistent with planar ground state structures, only if the torsional motion of lowest frequency occurs at about 80cm-1. It undergoes Heck reaction with 2-bromo-6-methoxynaphthalene in the presence of sodium acetate and Hermann′s catalyst in N,N-dimethylacetamide to afford diarylethene.

Application

4-Bromostyrene was used in the following studies:
  • Structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene.
  • Synthesis of silsesquioxanes (SQ) having 4-bromostyrenyl substituents.
  • To investigate the photochemical growth of Br-terminated self-assembled monolayers (SAMs) on Si(111).
  • Synthesis of poly(1,4-phenylenevinylene), via Heck reaction.
  • Synthesis of nitroolefins, via alkene cross-metathesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of monomers and polymers by the Heck reaction.
Heitz W, et al.
Makromol. Chem., 189(1), 119-127 (1988)
Synthesis and characterization of new hexahelicene derivatives.
Aloui F, et al.
Tetrahedron Letters, 48(11), 2017-2020 (2007)
Graham P Marsh et al.
Organic letters, 9(14), 2613-2616 (2007-06-07)
A synthesis of highly functionalized nitroalkenes is reported that utilizes a cross metathesis (CM) reaction between simple aliphatic nitro compounds and a range of substituted alkenes. This chemistry offers a simple and attractive route to nitroalkenes that would otherwise be
Małgorzata Bołt et al.
Materials (Basel, Switzerland), 13(18) (2020-09-12)
Bifunctional silsesquioxanes create an attractive group of compounds with a wide range of potential applications, and recently they have gained much interest. They are known to be obtained mainly via hydrosilylation, but we disclose novel synthetic protocols based on different
M Z Asuncion et al.
Journal of the American Chemical Society, 132(11), 3723-3736 (2010-03-02)
Polyphenylsilsesquioxane [PhSiO(1.5)](n) (PPS) and polyvinylsilsesquioxane [vinylSiO(1.5)](n) (PVS) are polymeric byproducts of the syntheses of the related T(8) octamers [PhSiO(1.5)](8) and [vinylSiO(1.5)](8). Here we demonstrate that random-structured PPS and PVS rearrange in the presence of catalytic amounts of Bu(4)N(+)F(-) in THF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service