523933
4-tert-Butylstyrene
contains ≤100 ppm tert-butylcatechol as inhibitor, 93%
Synonym(s):
1-tert -Butyl-4-vinylbenzene, p -tert -Butylstyrene
About This Item
Assay
93%
contains
≤100 ppm tert-butylcatechol as inhibitor
refractive index
n20/D 1.526 (lit.)
bp
91-92 °C/9 mmHg (lit.)
density
0.875 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(C)(C)c1ccc(C=C)cc1
InChI
1S/C12H16/c1-5-10-6-8-11(9-7-10)12(2,3)4/h5-9H,1H2,2-4H3
InChI key
QEDJMOONZLUIMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
177.8 °F - closed cup
Flash Point(C)
81 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service