Skip to Content
Merck
All Photos(1)

Key Documents

124060

Sigma-Aldrich

2-Bromofluorene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9Br
CAS Number:
Molecular Weight:
245.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

bp

185 °C/135 mmHg (lit.)

mp

112-114 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C13H9Br/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7H2

InChI key

FXSCJZNMWILAJO-UHFFFAOYSA-N

General description

2-Bromofluorene is a nonintrusive end-capping reagent to control molecular weights and to generate well-defined oligomers.

Application

2-Bromofluorene was used in the synthesis of end-capping reagent 2-bromo-9,9-di-nhexylfluorene. It has been used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization. It was also used in the synthesis of 2-bromo-9,9-dihexylfluorene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyfluorene derivatives: effective conjugation lengths from well-defined oligomers.
Klaerner G and Miller RD.
Macromolecules, 31(6), 2007-2009 (1998)
Tetrahedron Letters, 48, 89-89 (2007)
Well-defined [3, 3'] bicarbazolyl-based electroactive compounds for optoelectronics.
Vaitkeviciene V, et al.
Synthetic Metals, 158(8), 383-390 (2008)
Excimer and Aggregate Formations in Poly (fluorene) s.
Lee JI, et al.
ETRI Journal, 24(6), 409-414 (2002)
Tetrahedron Letters, 47, 7085-7085 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service