Skip to Content
Merck
All Photos(1)

Key Documents

123714

Sigma-Aldrich

4-(Benzyloxy)benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CHO
CAS Number:
Molecular Weight:
212.24
Beilstein:
1242385
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Quality Level

Assay

97%

mp

71-74 °C (lit.)

functional group

aldehyde
phenyl

SMILES string

[H]C(=O)c1ccc(OCc2ccccc2)cc1

InChI

1S/C14H12O2/c15-10-12-6-8-14(9-7-12)16-11-13-4-2-1-3-5-13/h1-10H,11H2

InChI key

ZVTWZSXLLMNMQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Benzyloxy)benzaldehyde was used in the synthesis of (5-fluoro-(2R*,3S*)-2,3-bis(4-hydroxyphenyl)pentanenitrile),an estrogen receptor β-selective ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jeongsoo Yoo et al.
Journal of medicinal chemistry, 48(20), 6366-6378 (2005-09-30)
Estrogen receptor beta (ERbeta), a less active ER subtype that appears to have a restraining effect on the more active ERalpha, could be a factor that determines the level of estrogen action in certain estrogen target tissues. ERbeta is found
Oriol Penon et al.
Journal of colloid and interface science, 462, 154-165 (2015-10-12)
The preparation of novel porphyrin derivatives and their immobilization onto iron oxide nanoparticles to build up suitable nanotools for potential use in photodynamic therapy (PDT) has been explored. To achieve this purpose, a zinc porphyrin derivative, ZnPR-COOH, has been synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service