Skip to Content
Merck
All Photos(1)

Key Documents

122432

Sigma-Aldrich

4-Amino-1-naphthalenecarbonitrile

97%

Synonym(s):

4-Cyano-1-naphthylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6CN
CAS Number:
Molecular Weight:
168.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

174.5-176.5 °C (lit.)

SMILES string

Nc1ccc(C#N)c2ccccc12

InChI

1S/C11H8N2/c12-7-8-5-6-11(13)10-4-2-1-3-9(8)10/h1-6H,13H2

InChI key

LUSIZUFVMKYWGX-UHFFFAOYSA-N

General description

4-Amino-1-naphthalenecarbonitrile on hydrolysis yields 4-amino-1-naphthoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Qian et al.
Bioorganic & medicinal chemistry, 7(12), 3011-3024 (2000-02-05)
Cysteine farnesylation at the carboxylate terminal tetrapeptide CAAX of Ras protein is catalyzed by farnesyltransferase. This lipid modification is necessary for regulatory function of both normal and oncogenic Ras. The high frequency of Ras mutation in human cancers has prompted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service