Skip to Content
Merck
All Photos(2)

Key Documents

117684

Sigma-Aldrich

2-(N-Ethylanilino)ethanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(C2H5)CH2CH2OH
CAS Number:
Molecular Weight:
165.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

268 °C (lit.)

mp

36-38 °C (lit.)

SMILES string

CCN(CCO)c1ccccc1

InChI

1S/C10H15NO/c1-2-11(8-9-12)10-6-4-3-5-7-10/h3-7,12H,2,8-9H2,1H3

InChI key

HYVGFUIWHXLVNV-UHFFFAOYSA-N

General description

2-(N-Ethylanilino) ethanol couples with 2-amino-5-formylthiophene during the synthesis of nonlinear optical chromophores. It undergoes azo coupling with 2-cyan-4-nitroaniline to yield azochromophore AZ1.

Application

2-(N-Ethylanilino) ethanol was used in the preparation of azo compound N-ethyl-N-(2-hydroxylethyl)-4-(2-cyano-4-nitrophenylazo) aniline (AZ1)2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photonics Letters of Poland null
Electro-optic guest-host system containing azobenzene chromophores in PMMA matrix.
Kozanecka-Szmigiel A, et al.
Photonics Letters of Poland, 1(4), 181-181 (2009)
Properties of novel azodyes containing powerful acceptor groups and thiophene moiety.
Ledoux I, et al.
Synthetic Metals, 115(1), 213-217 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service