Skip to Content
Merck
All Photos(1)

Key Documents

115819

Sigma-Aldrich

2,2′-Biphenol

99%

Synonym(s):

2,2′-Biphenyldiol, 2,2′-Dihydroxybiphenyl, 2,2′-Diphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4C6H4OH
CAS Number:
Molecular Weight:
186.21
Beilstein:
1638363
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

solid

bp

315 °C (lit.)

mp

108-110 °C (lit.)

SMILES string

Oc1ccccc1-c2ccccc2O

InChI

1S/C12H10O2/c13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14/h1-8,13-14H

InChI key

IMHDGJOMLMDPJN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

309.2 °F - closed cup - (External MSDS)

Flash Point(C)

154 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernd Schmidt et al.
The Journal of organic chemistry, 78(17), 8680-8688 (2013-08-01)
User-friendly protocols for the protecting group-free synthesis of 2,2'-biphenols via Suzuki-Miyaura coupling of o-halophenols and o-boronophenol are presented. The reactions proceed in water in the presence of simple additives such as K2CO3, KOH, KF, or TBAF and with commercially available
R J Heath et al.
The Journal of biological chemistry, 273(46), 30316-30320 (1998-11-07)
The broad spectrum antibacterial properties of 2-hydroxydiphenyl ethers have been appreciated for decades, and their use in consumer products is rapidly increasing. We identify the enoyl-acyl carrier protein reductase (fabI) component of the type II fatty acid synthase system as
Sierra Rayne et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 4(11), 876-886 (2005-10-28)
Photochemical studies on a range of model dibenzo[1,4]dioxins were performed in aqueous and organic solutions. The compounds were found to undergo a photochemically initiated aryl-ether bond homolysis that yields reactive 2-spiro-6'-cyclohexa-2',4'-dien-1'-one and subsequent 2,2'-biphenylquinone intermediates. Under steady-state irradiation, the 2,2'-biphenylquinones
Christina DiMarco-Crook et al.
Journal of food science, 85(4), 1292-1301 (2020-03-08)
Chemoprevention strategies employing the use of multiple dietary bioactive components and their metabolites in combination offer advantages due to their low toxicity and potential synergistic interactions. Herein, for the first time, we studied the combination of curcumin and 3',4'-didemethylnobiletin (DDMN)
T Kuhnigk et al.
Journal of basic microbiology, 37(3), 205-211 (1997-01-01)
The capability of the intestinal flora from the gut of xylophagous termites of degrading lignin model compounds was investigated. Different dimeric lignin model compounds-degrading bacteria were obtained from the hindgut flora of Mastotermes darwiniensis Froggatt, Reticulitermes santonensis Feytaud, Nasutitermes nigriceps

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service