Skip to Content
Merck
All Photos(1)

Key Documents

113727

Sigma-Aldrich

Norphenylephrine hydrochloride

98%

Synonym(s):

α-Aminomethyl-3-hydroxybenzyl alcohol hydrochloride, 3,α-Dihydroxyphenethylamine hydrochloride, Norfenefrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(CH2NH2)OH · HCl
CAS Number:
Molecular Weight:
189.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

162-164 °C (lit.)

functional group

amine
hydroxyl

SMILES string

Cl.NCC(O)c1cccc(O)c1

InChI

1S/C8H11NO2.ClH/c9-5-8(11)6-2-1-3-7(10)4-6;/h1-4,8,10-11H,5,9H2;1H

InChI key

OWMFSWZUAWKDRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Norphenylephrine hydrochloride was used in the study of the effects of phenylephrine on the isolated heart of rabbit.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Oldenburg et al.
Deutsche medizinische Wochenschrift (1946), 124(1-2), 8-12 (1999-02-10)
A 71-year-old man was admitted because of treatment-resistant orthostatic hypotension of unknown aetiology. When aged 64 years he developed some impotence and later urinary incontinence and urinary frequency. At 68 years he noted vertigo on physical activity, and a year
J F Coulon et al.
Journal of neurochemistry, 52(5), 1418-1424 (1989-05-01)
Phenolamines, particularly octopamines, are of special importance in avoidance behavior. In the Roman low avoidance (RLA) strain, p-octopamine can induce locomotor behavioral activity that is normally observed in the Roman high avoidance (RHA) strain. For these reasons, the levels of
M R Gaab et al.
Advances in neurology, 52, 391-400 (1990-01-01)
We investigated the effects of the calcium entry blocker nimodipine on cerebral autoregulation, BBB, and vasogenic edema in animal experiments. In the first series of rats, ICBF was measured with H2 clearance using a balanced multiwire surface electrode. Variations in
N Schwick et al.
Zeitschrift fur Kardiologie, 86(3), 165-170 (1997-03-01)
Measurement of baroreflex sensitivity is a new method to identify patients after myocardial infarction with a high risk for sudden cardiac death, ventricular tachycardia or ventricular fibrillation. In this retrospective study the baroreflex sensitivity was obtained noninvasively by measuring the
M Hennersdorf et al.
Zeitschrift fur Kardiologie, 86(3), 196-203 (1997-03-01)
To evaluate patients with an increased risk of sudden cardiac death the analysis of ventricular late potentials, heart rate variability and baroreflexsensitivity is helpful. However, the prediction of malignant arrhythmic events cannot be performed with sufficient accuracy. For a better

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service