Skip to Content
Merck
All Photos(3)

Key Documents

112968

Sigma-Aldrich

Methylhydroquinone

99%

Synonym(s):

Toluhydroquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
124.14
Beilstein:
2041489
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

autoignition temp.

851 °F

mp

128-130 °C (lit.)

SMILES string

Cc1cc(O)ccc1O

InChI

1S/C7H8O2/c1-5-4-6(8)2-3-7(5)9/h2-4,8-9H,1H3

InChI key

CNHDIAIOKMXOLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylhydroquinone is produced by the oxidation of o-cresol by the mutants G103S, G103S/A107G, and G103S/A107T.

Application

Methylhydroquinone can be used as a reactant to prepare:
  • A semiflexible thermotropic polyester via polycondensation reaction with 4,4′-sebacoyldioxydibenzoyl chloride.
  • A sesquiterpene (±)-helibisabonol A.
  • poly{hexakis[(methyl)(4-hydroxyphenoxy)]cyclotriphosphazene} by reacting with hexachlorocyclotriphosphazene.
  • 6-Hydroxy-4,7-dimethyl-2H-1-benzopyran-2-one by treating with ethyl acetoacetate in the presence of H2SO4 as a catalyst.

Biochem/physiol Actions

Methylhydroquinone (Toluquinol) inhibits the growth of endothelial and tumor cells in culture in the micromolar range and is a promising drug candidate in the treatment of cancer and other angiogenesis-related pathologies.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

341.6 °F - closed cup

Flash Point(C)

172 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and mesomorphic properties of a semiflexible polyester based on 4, 4?-sebacoyldioxydibenzoyl chloride and methylhydroquinone
Costa Giovanna, et al.
Macromolecular Chemistry and Physics, 191(4), 791-800 (1990)
Synthesis and characterization of poly {hexakis [(methyl)(4-hydroxyphenoxy)] cyclotriphosphazene}
Luther T A, et al.
Journal of Applied Polymer Science, 82(14), 3439-3446 (2001)
Solvent-Free Synthesis and Insecticidal Activities of C6-Esterified Coumarins
Xie Ying, et al.
Letters in Drug Design & Discovery, 11(9), 1124-1132 (2014)
First total synthesis of ?-helibisabonol A
Macias F A, et al.
Tetrahedron Letters, 43(36), 6417-6420 (2002)
Ying Tao et al.
Journal of bacteriology, 186(14), 4705-4713 (2004-07-03)
Wild-type toluene 4-monooxygenase (T4MO) of Pseudomonas mendocina KR1 oxidizes toluene to p-cresol (96%) and oxidizes benzene sequentially to phenol, to catechol, and to 1,2,3-trihydroxybenzene. In this study T4MO was found to oxidize o-cresol to 3-methylcatechol (91%) and methylhydroquinone (9%), to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service