Skip to Content
Merck
All Photos(1)

Key Documents

8.21233

Sigma-Aldrich

4-Methoxyphenol

for synthesis

Synonym(s):

4-Methoxyphenol, 1-Hyroxy-4-methoxybenzene, Hydroquinone monomethyl ether, MEHQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
205-769-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.009 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

421 °C

potency

1600 mg/kg LD50, oral (Rat)

pH

5.1 (20 °C, 30 g/L in H2O)

bp

234-246 °C/1013 hPa

mp

54-57 °C

transition temp

flash point 131 °C

solubility

40 g/L

density

1.55 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)c1ccc(cc1)O

InChI

1S/C7H8O2/c1-9-7-4-2-6(8)3-5-7/h2-5,8H,1H3

InChI key

NWVVVBRKAWDGAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Development of electrochemical immunosensors: 4-Methoxyphenol, is used in developing highly sensitive electrochemical immunosensors due to its favorable electrochemical properties, highlighting its biomedical application in sensor technology (Park et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 57 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service