Skip to Content
Merck
All Photos(1)

Key Documents

8.02892

Sigma-Aldrich

Dibutyl ether

for synthesis

Synonym(s):

Dibutyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2O
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352112
EC Index Number:
205-575-3
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
140-143 °C/1013 hPa
vapor pressure:
6.4 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

6.4 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

175 °C

potency

3200 mg/kg LD50, oral (Rat)
7640 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-8.5 % (v/v)

dilution

(for synthesis)

pH

5.2 (20 °C, 0.2 g/L in H2O)

bp

140-143 °C/1013 hPa

mp

-95 °C

transition temp

flash point 25 °C

solubility

10 g/L

density

0.769 g/cm3 at 20 °C

SMILES string

O(CCCC)CCCC

InChI

1S/C8H18O/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

DURPTKYDGMDSBL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99,0 Area %
Density (d 20 °C/ 4 °C): 0,767 - 0,769
Peroxide (as H₂O₂): ≤ 0,05 %(m)
Identity (IR): conforms

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service