Skip to Content
Merck
All Photos(1)

Key Documents

10907

Sigma-Aldrich

Chloroacetaldehyde solution

Wacker Chemie AG, ≥45.0% in H2O (density determination)

Synonym(s):

CLACH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHO
CAS Number:
Molecular Weight:
78.50
Beilstein:
1071226
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

manufacturer/tradename

Wacker Chemie AG

concentration

≥45.0% in H2O (density determination)

bp

80-100 °C (lit.)

functional group

aldehyde
chloro

SMILES string

[H]C(=O)CCl

InChI

1S/C2H3ClO/c3-1-2-4/h2H,1H2

InChI key

QSKPIOLLBIHNAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chloroacetaldehyde is a reagent used for converting adenine and its nucleoseides into fluorescent etheno derivatives which can be analysed by HPLC. It was used for derivatization in fluorometric determination of arprinocid and analogous compounds in human plasma .

Biochem/physiol Actions

Chloroacetaldehyde, a metabolite of ifosamide, influences oxidative phosphorylation in mitochondria. It causes breaking of DNA strands and strongly inhibits DNA synthesis.

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorogenic reaction between adenine derivatives and chloroacetaldehyde and its application to the determination of 9-(2-chloro-6-fluorobenzyl) adenine in human plasma.
Matuszewski BK and Bayne WF.
Analytica Chimica Acta, 227, 189-202 (1989)
Preparation, titration, and storage of chloroacetaldehyde for fluorometric determination of adenine and its derivatives.
McCann WP, et al.
Analytical Chemistry, 55(8), 1454-1455 (1983)
Svenja K Brüggemann et al.
Cancer chemotherapy and pharmacology, 57(3), 349-356 (2005-09-01)
The ifosfamide metabolite chloroacetaldehyde had been made responsible for side effects only. We found in previous studies a strong cytotoxicity on human MX-1 tumor cells and xenografts in nude mice. Chloroacetaldehyde is supposed to act via alkylation or by inhibition
Min Young Kim et al.
Chemical research in toxicology, 20(8), 1075-1083 (2007-07-31)
2-Chloroacetaldehyde (CAA), a metabolite of the carcinogen vinyl chloride, reacts with DNA to form cyclic etheno ()-lesions. AlkB, an iron-/alpha-ketoglutarate-dependent dioxygenase, repairs 1, N (6)-ethenodeoxyadenosine (A) and 3, N (4)-ethenodeoxycytidine (C) in site-specifically modified single-stranded viral genomes in vivo and
Leila Choucha-Snouber et al.
Biotechnology and bioengineering, 110(2), 597-608 (2012-08-14)
In this article, we present a liver-kidney co-culture model in a micro fluidic biochip. The liver was modeled using HepG2/C3a and HepaRG cell lines and the kidney using MDCK cell lines. To demonstrate the synergic interaction between both organs, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service