Skip to Content
Merck

C19406

Chloroacetaldehyde dimethyl acetal

98%

Synonym(s):

2-Chloro-1,1-dimethoxyethane, Dimethyl chloroacetal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
ClCH2CH(OCH3)2
CAS Number:
Molecular Weight:
124.57
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
202-624-0
Beilstein/REAXYS Number:
1733700
MDL number:
Assay:
98%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

128-130 °C (lit.)

density

1.094 g/mL at 25 °C (lit.)

SMILES string

COC(CCl)OC

InChI

1S/C4H9ClO2/c1-6-4(3-5)7-2/h4H,3H2,1-2H3

InChI key

CRZJPEIBPQWDGJ-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Chloroacetaldehyde dimethyl acetal


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

84.2 °F - closed cup

flash_point_c

29 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library