Skip to Content
Merck
All Photos(1)

Key Documents

10905

Sigma-Aldrich

Chloroacetone

Wacker Chemie AG, ≥96.0% (GC)

Synonym(s):

MCA, 1-Chloropropan-2-one, Chloro-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COCH3
CAS Number:
Molecular Weight:
92.52
Beilstein:
605369
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (GC)

contains

~0.1% Drapex 39 as stabilizer

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.432 (lit.)

bp

120 °C (lit.)

solubility

H2O: soluble 10 parts
alcohol: miscible
chloroform: miscible
diethyl ether: miscible

density

1.162 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)CCl

InChI

1S/C3H5ClO/c1-3(5)2-4/h2H2,1H3

InChI key

BULLHNJGPPOUOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chloroacetone undergoes direct aldol reaction with 4-nitrobenzaldehyde catalyzed by L-proline amides. It reacts with carboxylic acids to form ester derivatives.

Application

Chloroacetone was used in microbial asymmetric reduction of chloracetone to synthesize chiral 1,2-epoxy propane. It was used in analysis of acids by gas chromatography. It was also used in the syntheis of methylthiazole derivatives by reacting with 1-((3-(naphtho[2,1-b]furan-2-yl)-1-phenyl-1H-pyrazol-4-yl) methylene)-2-phenylhydrazone.

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashraf H F Abd El-Wahab et al.
Molecules (Basel, Switzerland), 16(1), 307-318 (2011-01-27)
Vilsmeier formylation of 2-(1-phenylhydrazonoethyl)naphtho[2,1-b]furan (2) gave 3-naphtho[2,1-b]furan-2-yl-1-phenyl-1H-pyrazole-4-carbaldehyde (3), which was reacted with C- and N-nucleophiles to afford naphthofuranpyrazol derivatives 4-8. Treatment of 2-[(3-(naphtho[2,1-b]furan-2-yl)-1-phenyl-1H-pyrazol-4-yl)methylene]-malononitrile (4a) with reactants having active hydrogen and Et₃N gave the corresponding pyrazoline, pyran and chromene addition product
Analysis of carboxylic acids by gas chromatography. Derivatisation using chloroacetone.
McCalley DV, et al.
Chromatographia, 18(6), 309-312 (1984)
Synthesis of optically pure 1, 2-epoxypropane by microbial asymmetric reduction of chloroacetone.
Weijers CAGM, et al.
Applied Microbiology and Biotechnology, 38(3), 297-300 (1992)
L-Proline amide-catalyzed direct asymmetric aldol reaction of aldehydes with chloroacetone.
He L, et al.
Tetrahedron, 62(2), 346-351 (2006)
M R Hollaway et al.
The Biochemical journal, 191(3), 811-826 (1980-12-01)
1. Chloroacetone (I) was shown to be an active-site-directed inhibitor of the aliphatic amidase (EC 3.5.1.4) from Pseudomonas aeruginosa strain PAC142.2. This inhibitor reacted with the enzyme in two stages: the first involving the reversible formation of an enzymically inactive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service