Skip to Content
Merck
All Photos(1)

Key Documents

10896

Sigma-Aldrich

Vinyl laurate

Wacker Chemie AG, ≥98% (GC)

Synonym(s):

VL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COOCH=CH2
CAS Number:
Molecular Weight:
226.36
Beilstein:
1778369
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (GC)

manufacturer/tradename

Wacker Chemie AG

bp

254 °C/1013 hPa (lit.)

solubility

H2O: slightly soluble 1 g/L at 20 °C

density

0.871 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CCCCCCCCCCCC(=O)OC=C

InChI

1S/C14H26O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h4H,2-3,5-13H2,1H3

InChI key

GLVVKKSPKXTQRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vinyl laurate is a clear, colorless to light yellow colored liquid. The flash point is 125°C. It forms aqueous colloidal microgel by copolymerization with N-isopropylacrylamide. The enzymatic acylation of Avicel cellulose with vinyl laurate, a solvent free reaction system, was studied.

Application

Vinyl laurate was used in the synthesis of laurate and stearate esters of corn starch. It was used as the acyl donor during synthesis of sucrose laurate esters catalyzed by Bacillus pseudofirmus AL-89 . Vinyl laurate was also used in the preparation of chirally deuterated benzyl chlorides.

Other Notes

prices for bulk quantities on request

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

257.0 °F

Flash Point(C)

125 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of sucrose laurate using a new alkaline protease.
Pedersen NR, et al.
Tetrahedron Asymmetry, 14(6), 667-673 (2003)
Novel gelling behavior of poly (N-isopropylacrylamide-co-vinyl laurate) microgel dispersions.
Benee LS, et al.
Langmuir, 18(16), 6025-6030 (2002)
Synthesis of higher fatty acid starch esters using vinyl laurate and stearate as reactants.
Junistia L, et al.
Starch/Staerke, 60(12), 667-675 (2008)
Derek W Barnett et al.
Journal of labelled compounds & radiopharmaceuticals, 56(1), 6-11 (2013-11-29)
Chirally deuterated benzyl chlorides were prepared using novel, general hexachloroacetone/polymer-supported triphenylphosphine treatment of chirally deuterated benzyl alcohols. Doubly labeled protected tyrosine was obtained in 62% yield with 86% de at the α-carbon and 82% de at the β-carbon. Key in
Stavros Gremos et al.
Bioresource technology, 102(2), 1378-1382 (2010-10-05)
Cellulose esters are an important class of functional biopolymers with great interest in the chemical industry. In this work the enzymatic acylation of Avicel cellulose with vinyl propionate, vinyl laurate and vinyl stearate, has been performed successfully in a solvent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service