Skip to Content
Merck
All Photos(1)

Key Documents

638455

Sigma-Aldrich

Trifluoromethanesulfonamide

95%

Synonym(s):

1,1,1-Trifluoromethanesulfonamide, Trifluoromethylsulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO2NH2
CAS Number:
Molecular Weight:
149.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

120-124 °C (lit.)

SMILES string

NS(=O)(=O)C(F)(F)F

InChI

1S/CH2F3NO2S/c2-1(3,4)8(5,6)7/h(H2,5,6,7)

InChI key

KAKQVSNHTBLJCH-UHFFFAOYSA-N

Gene Information

human ... CA2(760)

Application

Trifluoromethanesulfonamide can undergo reaction with paraformaldehyde either in sulfuric acid to give the corresponding open chain and cyclic condensation products or in ethyl acetate to give the corresponding oxy-methylated products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Lynch et al.
The Biochemical journal, 310 ( Pt 1), 197-202 (1995-08-15)
The role of carbonic anhydrase in de novo lipid synthesis was examined by measuring [1-14C]acetate incorporation into total lipids, fatty acids and non-saponifiable lipids in freshly isolated rat hepatocytes. Two carbonic anhydrase inhibitors, trifluoromethylsulphonamide (TFMS) and ethoxozolamide (ETZ) decreased incorporation
"Oxymethylation of trifluoromethanesulfonamide with paraformaldehyde in ethyl acetate"
Meshcheryakov.I.V, et al.
Russ. J. Org. Chem., 44(02), 311-316 (2008)
"Cascade transformations of trifluoromethanesulfonamide in reaction with formaldehyde"
Meshcheryakov.I.V, et al.
Russ. J. Org. Chem., 41(09), 1381-1386 (2005)
Lina Baranauskienė et al.
BMC biophysics, 5, 12-12 (2012-06-09)
Human carbonic anhydrases (CAs) play crucial role in various physiological processes including carbon dioxide and hydrocarbon transport, acid homeostasis, biosynthetic reactions, and various pathological processes, especially tumor progression. Therefore, CAs are interesting targets for pharmaceutical research. The structure-activity relationships (SAR)
Marine Soulié et al.
Physical chemistry chemical physics : PCCP, 18(43), 29999-30008 (2016-11-03)
The arrangement of an ionic fluorophore in the crystalline state was regulated by the presence of various counter-ions and the effect on spectroscopic and self-association properties was studied. To do so, nine salts of berberine (i.e. a fluorescent natural alkaloid)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service