LC/MS Analysis of Anesthetics on Ascentis® Express F5
![LC/MS Analysis of Anesthetics on Ascentis® Express F5 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/237/068/c962bf0b-ceec-4c70-87d4-465b5ec1e688/800/c962bf0b-ceec-4c70-87d4-465b5ec1e688.jpg)
CONDITIONS
column
Ascentis Express F5, 10 cm x 4.6 mm I.D., 2.7 μm particles (53590-U)
mobile phase
[A] 10 mM ammonium acetate, pH 4.0 with acetic acid; [B] acetonitrile; (20:80, A:B)
flow rate
1.0 mL/min
pressure
2320 psi (160 bar)
column temp.
35 °C
detector
MS, ESI(+), SIR, m/z 235, 236, 247
injection
2 μL
sample
10 μg/mL in methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany