Merck
  • Amino acid-catalyzed formation of 2-vinylfuran from lipid-derived 4-oxo-2-hexenal.
Amino acid-catalyzed formation of 2-vinylfuran from lipid-derived 4-oxo-2-hexenal.
Food chemistry (2015-06-05)
Zhen Zeng, Jimei Ma, Bin Liu, Hong Jiang
ABSTRACT
The formation of 2-vinylfuran from the corresponding 4-oxo-2-hexenal (OHE, a lipid oxidation product) under the catalysis of amino acid were studied. The effects of amino acids, reaction temperature, reaction time, water content, pH, metallic ions and some food additives on the formation of 2-vinylfuran were investigated. Amino acids promoted the formation of 2-vinylfuran except aspartic acid, histidine and glutamine. Cysteine showed the strongest catalytic activity among the amino acids tested. Further investigation indicated that the intermediate 1-(furan-2-yl)ethanol played an very important role in the formation of 2-vinylfuran. Both higher temperature and acidic condition favored the transformation of OHE into 2-vinylfuran. The reaction time, water content, metallic ions and food additives also influenced the reaction to certain extent.
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
SAFC
Glycine
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)