Merck
Endophilin A2 Promotes TNBC Cell Invasion and Tumor Metastasis.
Molecular cancer research : MCR (2015-03-19)
Tomas Baldassarre, Kathleen Watt, Peter Truesdell, Jalna Meens, Mark M Schneider, Sandip K Sengupta, Andrew W Craig
ABSTRACT
Triple-negative breast cancers (TNBCs) are highly aggressive cancers that lack targeted therapies. However, EGFR is frequently activated in a subset of TNBCs and represents a viable clinical target. Because the endocytic adaptor protein Endophilin A2 (SH3GL1/Endo II) has been implicated in EGFR internalization, we investigated Endo II expression and function in human TNBCs. Endo II expression was high in several TNBC cells compared with normal breast epithelial cells. Stable knockdown (KD) of Endo II was achieved in two TNBC cell lines, and although cell viability was unaffected, defects in receptor-mediated endocytosis were observed. EGFR signaling to Erk and Akt kinases was impaired in Endo II KD cells, and this correlated with reduced rates of EGFR internalization and cell motility. Endo II KD cells also displayed defects in three dimensional (3D) cell invasion, and this correlated with impaired extracellular matrix degradation and internalization of MT1-MMP. Endo II silencing also caused a significant reduction in TNBC tumor growth and lung metastasis in mammary orthotopic tumor xenograft assays. In human breast tumor specimens, Endo II expression was highest in TNBC tumors compared with other subtypes, and at the level of gene expression, high Endo II was associated with reduced relapse-free survival in patients with basal-like breast cancers. Together, these results identify a positive role for Endo II in TNBC tumor metastasis and a potential link with poor prognosis. Endophilin A2 and related adaptor proteins represent important signaling hubs to target in metastatic cancers.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human CD74
Sigma-Aldrich
MISSION® esiRNA, targeting human GCNT2
Sigma-Aldrich
MISSION® esiRNA, targeting human TAF8
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Anti-SH3GL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human VIPR1