Merck
  • Monetite granules versus particulate autologous bone in bone regeneration.
Monetite granules versus particulate autologous bone in bone regeneration.
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft (2015-05-02)
Jesús Torres, Iskandar Tamimi, Jatsue Cabrejos-Azama, Isabel Tresguerres, Mohammad Alkhraisat, Enrique López-Cabarcos, Gonzalo Hernández, Faleh Tamimi
ABSTRACT
The aim of this study was to test bone tissue response to monetite granules in comparison with intramembranous autologous bone graft in a rabbit calvaria critical size defect model. Novel monetite granules were synthesized by thermal conversion of set brushite cement. Eight female New Zealand rabbits were used for this study. Two identical 10mm diameter bicortical cranial defects were created in each animal. One of the defects was grafted with monetite granules while the contralateral was grafted with granules of intramembranous autologous bone as control. Animals were sacrificed 8 weeks after surgery and biopsies were taken for histological and histomorphometrical evaluation under light microscopy. Wilcoxon test was used for statistical analysis. The bone defects treated with either autologous bone or monetite granules were able to heal within the study period. Upon histological observation the defects treated with autologous bone granules resembled the adjacent intact calvaria, whereas the defects treated with monetite showed a high infiltration of new bone and only 13.4±8.4% of remaining granules. The percentage of bone volume in the defects of the control group (71±9%) was 16% higher than in the study group (55±10%) (p<0.05). The percentage of augmented mineralized tissue volume in the study group (68±18%) was not significantly different from the control group (p>0.05). The amount of augmented mineralized tissue in the bone defects obtained with monetite granules was not significantly different from that obtained with autologous bone. This study confirms the potential of monetite based biomaterials as an alternative to autologous bone graft.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
4-Chloro-2-methylphenoxyacetic acid, technical, ≥95.0% (T)
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Dibasic Calcium Phosphate, meets USP testing specifications
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Calcium phosphate dibasic, 98.0-105.0%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis