Skip to Content
Merck
All Photos(1)

Documents

25190

Sigma-Aldrich

4-Chloro-2-methylphenoxyacetic acid

technical, ≥95.0% (T)

Synonym(s):

4-Chloro-o-tolyloxyacetic acid, MCPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)OCH2CO2H
CAS Number:
Molecular Weight:
200.62
Beilstein:
2051752
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥95.0% (T)

form

solid

mp

114-118 °C (lit.)

solubility

water: insoluble(lit.)

SMILES string

Cc1cc(Cl)ccc1OCC(O)=O

InChI

1S/C9H9ClO3/c1-6-4-7(10)2-3-8(6)13-5-9(11)12/h2-4H,5H2,1H3,(H,11,12)

InChI key

WHKUVVPPKQRRBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-2-methylphenoxyacetic acid (MPCA) is a herbicide. Various MCPA-mineralising bacterial combinations has been tested using a high-throughput microplate radiorespirometric mineralisation assay. Degradation mechanisms of OH and MCPA including molecular form and anionic form have been studied. Rapid and sensitive LC-electrospray tandem mass spectrometry method has been developed for the quantitation of MCPA in water and soil samples.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Urse S Krüger et al.
Pest management science, 71(2), 257-265 (2014-04-17)
The herbicide 4-chloro-2-methylphenoxyacetic acid (MCPA) is found frequently in Danish groundwater in concentrations exceeding the EU threshold limit of 0.1 µg L(-1) . Groundwater is used for drinking water, and one potential remediation strategy is bioaugmentation using inoculation of sand filters at
G G Bond et al.
British journal of industrial medicine, 50(4), 340-348 (1993-04-01)
For the purpose of assessing the human carcinogenic potential of the chlorophenoxy herbicides MCPA, MCPP, and 2,4-DP, the relevant epidemiological and toxicological evidence is reviewed. These compounds have not produced tumours in animal studies conducted under current test guidelines, giving
Edgar Hiller et al.
Chemosphere, 87(5), 437-444 (2011-12-31)
Herbicide leaching through soil into groundwater greatly depends upon sorption-desorption and degradation phenomena. Batch adsorption, desorption and degradation experiments were performed with acidic herbicide MCPA and three soil types collected from their respective soil horizons. MCPA was found to be
O Pozo et al.
Journal of chromatography. A, 923(1-2), 75-85 (2001-08-21)
A rapid and sensitive LC-electrospray tandem mass spectrometry method has been developed for the quantitation of 4-chloro-2-methylphenoxyacetic acid (MCPA) and 4-chloro-2-methylphenol in both water and soil samples. Soil samples were extracted in alkaline media and cleaned-up by solid-phase extraction with
Nora Badawi et al.
Environmental pollution (Barking, Essex : 1987), 170, 32-38 (2012-07-06)
Soil suspensions (slurries) are commonly used to estimate the potential of soil microbial communities to mineralize organic contaminants. The preparation of soil slurries disrupts soil structure, however, potentially affecting both the bacterial populations and their protozoan predators. We studied the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service