Merck
  • An analytical quality by design (aQbD) approach for a L-asparaginase activity method.
An analytical quality by design (aQbD) approach for a L-asparaginase activity method.
Journal of pharmaceutical and biomedical analysis (2015-09-18)
Han Yao, Jochem Vancoillie, Matthias D'Hondt, Evelien Wynendaele, Nathalie Bracke, Bart De Spiegeleer
ABSTRACT
L-asparaginase is an effective anti-tumor agent for acute lymphoblastic leukemia. This work presents the development of an activity determination of L-ASNase preparations for pharmaceutical quality control purposes, in accordance with analytical Quality by Design principles. Critical method attributes, the absorbance at 450 nm (A450) of the Nessler product as well as its variability, were evaluated as a function of critical method variables, by using experimental designs. The design space of the enzyme activity assay was defined (Nessler method: C(KI)/C(HgI2) of 1.90-1.95, C(NaOH)/C(HgI2) of 17.0-18.0, C(HgI2final) of 20-40 mM and time of 10-40 min; enzyme activity conditions: temperature range of 36.6-37.4 °C, pH range of the KH2PO4 buffer from 7.1 to 7.7, KH2PO4 buffer concentration: 0.18-0.22 M and L-Asn concentration of 18-22 mM), leading to a final enzyme activity assay method. A control strategy was ultimately implemented using system suitability tests.
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
8-Hydroxyquinoline, ACS reagent, 98.5%
Sigma-Aldrich
Citric acid monohydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, E330, 99.5-100.5% (based on anhydrous substance), grit
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
8-Hydroxyquinoline
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
L-Asparagine monohydrate, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Citric acid monohydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
L-Asparagine monohydrate, ≥99% (TLC)