Skip to Content
Merck
All Photos(4)

Key Documents

P9791

Sigma-Aldrich

Potassium phosphate monobasic

for molecular biology, ≥98.0%

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

for molecular biology

Assay

≥98.0%

form

powder

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

solubility

H2O: 100 mg/mL

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 ppm

cation traces

Fe: ≤20 ppm
heavy metals (as Pb): ≤10 ppm

application(s)

diagnostic assay manufacturing

foreign activity

DNase, Exonuclease; NICKase, Endonuclease; RNase and protease, none detected

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium phosphate monobasic has been used:
  • in high-throughput fluorescence assay of cytochrome P450 3A4[1]
  • in morphological analysis of neuromuscular junction development and degeneration in rodent lumbrical muscles[2]
  • in the measurement of net high-affinity urea uptake in maize plants[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of net high-affinity urea uptake in maize plants
Zanin L, et al.
BMC plant biology (2014)
Morphological analysis of neuromuscular junction development and degeneration in rodent lumbrical muscles
Sleigh J N, et al.
Journal of Neuroscience Methods, 227, 159-165 (2014)
High-throughput fluorescence assay of cytochrome P450 3A4
Cheng Q, et al.
Nature Protocols, 4(9), 1258-1258 (2009)
Przemyslaw Nogly et al.
IUCrJ, 2(Pt 2), 168-176 (2015-04-14)
Lipidic cubic phases (LCPs) have emerged as successful matrixes for the crystallization of membrane proteins. Moreover, the viscous LCP also provides a highly effective delivery medium for serial femtosecond crystallography (SFX) at X-ray free-electron lasers (XFELs). Here, the adaptation of
Casper Skovgaard et al.
Journal of applied physiology (Bethesda, Md. : 1985), 117(10), 1097-1109 (2014-09-06)
The purpose of this study was to examine whether speed endurance training (SET, repeated 30-s sprints) and heavy resistance training (HRT, 80-90% of 1 repetition maximum) performed in succession are compatible and lead to performance improvements in moderately trained endurance

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service