Merck
  • Proteomic Analysis of Estrogen-Mediated Signal Transduction in Osteoclasts Formation.
Proteomic Analysis of Estrogen-Mediated Signal Transduction in Osteoclasts Formation.
BioMed research international (2015-06-30)
Qi Xiong, Peifu Tang, Yanpan Gao, Lihai Zhang, Wei Ge
ABSTRACT
Estrogen plays an important role in inhibiting osteoclast differentiation and protecting against bone loss from osteoporosis, especially in postmenopausal women. However, the precise mechanisms underlying the effect of estrogen on osteoclasts are not well known. In the present study, we performed proteomics analysis and bioinformatics analysis to comprehensively compare the differential expression of proteins in receptor activator of nuclear factor-κB ligand RANKL-induced osteoclasts in the presence and absence of estrogen. We identified 6403 proteins, of which 124 were upregulated and 231 were downregulated by estrogen. Bioinformatics analysis showed that estrogen treatment interfered with 77 intracellular pathways, including both confirmed canonical and unconfirmed pathways of osteoclast formation. Our findings validate the inhibitory effect of estrogen on osteoclasts via the promotion of apoptosis and suppression of differentiation and polarization and suggest that estrogen might inhibit osteoclast formation via other pathways, which requires further investigation and verification.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Naphthol AS-BI Phosphoric Acid Solution
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)