Skip to Content
Merck
All Photos(2)

Key Documents

U1250

Sigma-Aldrich

Urea

ReagentPlus®, ≥99.5%, pellets

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99.5%

form

pellets

mp

132-135 °C (lit.)

solubility

H2O: 8 M

density

1.335 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Urea is a chaotropic agent and is used for protein denaturation. It disturbs the hydrogen bonds in the secondary, tertiary and quaternary structure of proteins. Urea can also disturb hydrogen bonds present in DNA secondary structure.

Application

Urea has been used as a protein denaturing agent.
Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Sheehan D.
Physical Biochemistry: Principles and Applications (2013)
Protein carbonyl determination by a rhodamine B hydrazide-based fluorometric assay.
Georgiou C D, et al.
Redox Biology, 17, 236-245 (2018)
Protein and cell wall polysaccharide carbonyl determination by a neutral pH 2, 4-dinitrophenylhydrazine-based photometric assay.
Georgiou C D, et al.
Redox Biology, 17, 128-142 (2018)
Christos D Georgiou et al.
Redox biology, 17, 128-142 (2018-04-24)
A new 2,4-dinitrophenylhydrazine (DNPH)-based photometric assay is developed for the quantification of carbonyls in protein samples from any biological source by protein carbonyl-DNPH hydrazone formation at acidic pH in the presence of denaturing urea, and subsequent hydrazone solubilization in the
Sreenivasa C Ramaiahgari et al.
Archives of toxicology, 88(5), 1083-1095 (2014-03-07)
Immortalized hepatocyte cell lines show only a weak resemblance to primary hepatocytes in terms of gene expression and function, limiting their value in predicting drug-induced liver injury (DILI). Furthermore, primary hepatocytes cultured on two-dimensional tissue culture plastic surfaces rapidly dedifferentiate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service