Merck
  • Protein arginine methyltransferase 10 is required for androgen-dependent proliferation of LNCaP prostate cancer cells.
Protein arginine methyltransferase 10 is required for androgen-dependent proliferation of LNCaP prostate cancer cells.
Bioscience, biotechnology, and biochemistry (2015-03-24)
Naoki Harada, Toshiki Takagi, Yoshihisa Nakano, Ryoichi Yamaji, Hiroshi Inui
ABSTRACT
Androgen receptor (AR) signaling is the master regulator of prostate cell growth. Here, to better understand AR signaling, we searched for AR-interacting proteins by yeast two-hybrid screening and identified protein arginine methyltransferase 10 (PRMT10) as one of the interacting proteins. PRMT10 was highly expressed in reproductive tissues, such as prostate. Immunostaining showed that PRMT10 was expressed in the nucleus of both epithelia and stroma of rat prostate. In human prostate cancer LNCaP cells, PRMT10 co-immunoprecipitated with AR in both the presence and absence of dihydrotestosterone (DHT). Knockdown of PRMT10 by siRNA decreased DHT-dependent LNCaP cell growth and induction of prostate-specific antigen, an AR-target gene, without apparent loss of AR. DHT decreased PRMT10 at both the mRNA and protein levels. The decrease in PRMT10 was canceled by knockdown of AR or an AR antagonist. These results indicate that PRMT10 plays an important role in androgen-dependent proliferation of prostate cancer cells.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Anti-PRMT9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis