Merck
  • Enhanced expression of matrix metalloproteinase-12 contributes to Npc1 deficiency-induced axonal degeneration.
Enhanced expression of matrix metalloproteinase-12 contributes to Npc1 deficiency-induced axonal degeneration.
Experimental neurology (2015-04-14)
Guanghong Liao, Zhuangjun Wang, Erik Lee, Stephanie Moreno, Omar Abuelnasr, Michel Baudry, Xiaoning Bi
ABSTRACT
Niemann-Pick type C (NPC) disease is a genetic disorder associated with intracellular cholesterol accumulation in the brain and other organs, and neurodegeneration is generally believed to be the fatal cause of the disease. In view of the emerging role of matrix metalloproteinase-12 (MMP-12) in neuronal injury, we investigated its expression and potential roles in axonal degeneration in Npc1-/- mouse brain. Microarray and quantitative real-time reversed transcription PCR analysis indicated a marked increase in MMP-12 mRNA levels in cerebellum of 3 week-old Npc1-/- mice, as compared to wild-type littermates. Western blots showed that the ratio of mature MMP-12 over pro-MMP-12 was significantly increased in cerebellum of Npc1-/-, as compared to wild-type mice. Immunohistochemical studies confirmed that MMP-12 expression was increased, especially in the cell bodies of Purkinje neurons in Npc1-/- mice. Neuritic growth was significantly reduced by Npc1 siRNA knockdown in nerve growth factor-differentiated PC-12 cells, and this effect was completely reversed by treatment with an MMP-12 specific inhibitor. Furthermore, in vivo experiments showed that chronic treatment with the MMP-12 inhibitor ameliorated Npc1 deficiency-induced axonal pathology in the striatum. Our results indicate that abnormal neuronal expression of MMP-12 may contribute to axonal degeneration in NPC disease, thus providing a potential novel target for treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human NPC1
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt ≥300,000
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Npc1
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)