Merck
  • Compact and blinking-suppressed quantum dots for single-particle tracking in live cells.
Compact and blinking-suppressed quantum dots for single-particle tracking in live cells.
The journal of physical chemistry. B (2014-08-27)
Lucas A Lane, Andrew M Smith, Tianquan Lian, Shuming Nie
ABSTRACT
Quantum dots (QDs) offer distinct advantages over organic dyes and fluorescent proteins for biological imaging applications because of their brightness, photostability, and tunability. However, a major limitation is that single QDs emit fluorescent light in an intermittent on-and-off fashion called "blinking". Here we report the development of blinking-suppressed, relatively compact QDs that are able to maintain their favorable optical properties in aqueous solution. Specifically, we show that a linearly graded alloy shell can be grown on a small CdSe core via a precisely controlled layer-by-layer process, and that this graded shell leads to a dramatic suppression of QD blinking in both organic solvents and water. A substantial portion (>25%) of the resulting QDs does not blink (more than 99% of the time in the bright or "on" state). Theoretical modeling studies indicate that this type of linearly graded shell not only can minimize charge carrier access to surface traps but also can reduce lattice defects, both of which are believed to be responsible for carrier trapping and QD blinking. Further, we have evaluated the biological utility of blinking-suppressed QDs coated with polyethylene glycol (PEG)-based ligands and multidentate ligands. The results demonstrate that their optical properties are largely independent of surface coatings and solvating media, and that the blinking-suppressed QDs can provide continuous trajectories in live-cell receptor tracking studies.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Supelco
Bis(2-ethylhexyl) adipate, 99%
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥97.0% (GC)
Supelco
Bis(2-ethylhexyl) adipate, Selectophore, ≥99.0%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%