Merck
  • A simple multi-residue method for the determination of pesticides in fruits and vegetables using a methanolic extraction and ultra-high-performance liquid chromatography-tandem mass spectrometry: optimization and extension of scope.
A simple multi-residue method for the determination of pesticides in fruits and vegetables using a methanolic extraction and ultra-high-performance liquid chromatography-tandem mass spectrometry: optimization and extension of scope.
Journal of chromatography. A (2015-02-11)
V Hanot, S Goscinny, M Deridder
ABSTRACT
In 2004, a new multi-residue pesticides method had been published using methanol as extraction solvent. Our goal for this study was to optimize the analytical scheme while extending the compound scope from 19 to 200 pesticides. The main changes from the original method take place at the sample extraction and processing with a special attention to make the overall method fit for routine analysis with minimal cost. Hence, after a quick Ultra-Turrax homogenization with a methanolic solution, the sample is simply diluted before the separation and detection by ultra-high-performance liquid chromatography and MS/MS detection for quantitative and confirmatory purposes. The performance of the method including limits of quantification (LOQs), linearity, matrix effect, precision was evaluated during validation in accordance with the European Union SANCO/12571/2013 regulatory guidelines. Two representative matrices, lettuce and orange, were selected and fortified at two concentration levels for these experiments. At the LOQ and ten times the LOQ, recoveries of the analytes were mostly within 70-120%, with coefficients of variation lower than 25% in intra-day repeatability conditions. In addition to being simple and fast, these results demonstrate the suitability of the optimized method for the analysis of large scope pesticides in routine laboratories.
Product Description
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Chlorosulfuron, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Cyazofamid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis