Merck
  • Residues investigation of fluoroquinolones and sulphonamides and their metabolites in bovine milk by quantification and confirmation using ultra-performance liquid chromatography-tandem mass spectrometry.
Residues investigation of fluoroquinolones and sulphonamides and their metabolites in bovine milk by quantification and confirmation using ultra-performance liquid chromatography-tandem mass spectrometry.
Food chemistry (2014-12-23)
Zhe Meng, Zhihong Shi, Shuxuan Liang, Xinfeng Dong, Hui Li, Hanwen Sun
ABSTRACT
A novel UPLC-MS/MS method for quantification and confirmation of eight fluoroquinolones, five sulphonamides (SAs) and four acetyled metabolites in milk is developed. Their main fragmentation pathways were presented. The limits of quantification were in the range of 0.01-0.29 μg kg(-1) for FQs and 0.13-1.68 μg kg(-1) for SAs. Mean recoveries ranging from 61% to 115% were achieved at spiked level of 0.5-100 μg kg(-1). The intra- and inter-day precisions were in the range of 6.3-10.7% and 9.0-13.4%, respectively. This novel approach has high speed and sensitivity, and can ensure the multiclass and multiresidue analysis of bovine milk at low μg kg(-1) level. The content of each analyte residue is much lower than the maximum residue limits. Some residues in milk were confirmed in according to the Commission Decision 657/2002/EC.
Product Description
Supelco
Sulfamonomethoxine, VETRANAL®, analytical standard
Supelco
Sulfadimethoxine, 98.0-102.0%
Sigma-Aldrich
Sulfamonomethoxine
Sigma-Aldrich
Sulfameter
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Norfloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sulfadimethoxine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfadimethoxine, Pharmaceutical Secondary Standard; Certified Reference Material
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sulfadimidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfadiazine for identification of impurity F, European Pharmacopoeia (EP) Reference Standard
Norfloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Supelco
Sulfamerazine, VETRANAL®, analytical standard
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Sulfamerazine, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Sulfamethoxazole
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Norfloxacin, VETRANAL®, analytical standard