Skip to Content
Merck
All Photos(1)

Key Documents

S7007

Supelco

Sulfadimethoxine

98.0-102.0%

Synonym(s):

4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Beilstein:
306856
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

98.0-102.0%

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

forensics and toxicology
pharmaceutical (small molecule)

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

COc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC)n1

InChI

1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ZZORFUFYDOWNEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Sulfonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase.
Mode of Action: Inhibits folic acid synthesis in prokaryotes.
Anti-microbial Spectrum: Gram positive, Gram negative, Chlamydia
Mode of Resistance: Alteration of dihydropteroate synthase or alternative pathway for folic acid synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Annie von Eyken et al.
Journal of the American Society for Mass Spectrometry, 30(5), 765-777 (2019-03-17)
Non-targeted screening (e.g., suspected-target) is emerging as an attractive tool to investigate the occurrence of contaminants in food. The sample preparation and instrument analysis steps are known to influence the identification of analytes with non-targeted workflows, especially for complex matrices.
Monica J Daood et al.
Pediatric research, 72(5), 455-459 (2012-08-21)
Hazardous levels of bilirubin produce oxidative stress in vitro and may play a role in the genesis of bilirubin-induced neurologic dysfunction (BIND). We hypothesized that the antioxidants taurourosdeoxycholic acid (TUDCA), 12S-hydroxy-1,12-pyrazolinominocycline (PMIN), and minocycline (MNC) inhibit oxidative stress and block
Hiroichi Ozaki et al.
Veterinary microbiology, 150(1-2), 132-139 (2011-01-15)
To investigate the effects of rearing practices of commercial broiler chickens on the incidence of antimicrobial resistance in commensal Escherichia coli isolates, fecal E. coli isolates obtained in 4 farms were screened for anitimicrobial resistance. Ten E. coli isolates were
Dan Liu et al.
Chemical & pharmaceutical bulletin, 59(1), 63-71 (2011-01-08)
Recently, dendrimers have been widely used in medical applications such as drug delivery and gene transfection. In this study, a pH-sensitive diblock copolymer of poly(methacryloyl sulfadimethoxine) (PSD) and polyethylene glycol (PEG) modified by lactose (LA-PEG-b-PSD) was synthesized. The pK(a) value
Ailiang Chen et al.
Biosensors & bioelectronics, 42, 419-425 (2012-12-12)
Quickly and sensitively detection of antibiotic residues in animal products often requires time-consuming techniques and expensive instrumentation. To address these limitations, a high sensitive aptasensor for sulfadimethoxine (SDM) using unmodified gold nanoparticles (AuNPs) was developed in the present study. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service