Merck
  • Development of ciclopirox olamine topical formulations: evaluation of drug release, penetration and cutaneous retention.
Development of ciclopirox olamine topical formulations: evaluation of drug release, penetration and cutaneous retention.
Pharmaceutical development and technology (2013-11-30)
Vanessa Alves Pinheiro, Daniela Serikaku, Andre Rolim Baby, Maria Valéria Robles Velasco, Telma Mary Kaneko, Vladi Olga Consiglieri
ABSTRACT
With the aim of reducing system absorption and consequently, the side effects, and simultaneously select a penetration enhancing, three topical formulations with 0.5% ciclopirox olamine (CO) and 15% of propylene glycol (PG), ethoxydiglycol or oleic acid were developed and evaluated regarding the skin penetration and cutaneous retention of the drug using Franz diffusion cells. Release experiments were performed through synthetic membrane while dermatomed pig ear skin was used to evaluate CO skin penetration and skin retention. Retention studies were carried out applying tape stripping method and dosing CO in stratum corneum and in epidermis and dermis. A HPLC method was validated for quantifying CO. All formulations tested with synthetic membrane presented no retention of the drug. Permeation data suggested that there was no systemic absorption of ciclopirox olamine from the studied formulations, even when the skin penetration enhancers were applied. Higher concentrations of the drug were found in the stratum corneum (SC) and also in epidermis and dermis, for all of the developed formulations. The addition of enhancers improved the penetration and cutaneous retention of CO, and propylene glycol promoted higher concentrations in epidermis and dermis, probably because its cumulative effect on the skin and by an efficient solvent power.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, analytical standard
Supelco
Octyldodecanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ciclopirox olamine, United States Pharmacopeia (USP) Reference Standard
USP
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Ciclopirox olamine, European Pharmacopoeia (EP) Reference Standard
Octyldodecanol, European Pharmacopoeia (EP) Reference Standard
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%