Merck
  • Enzymatic characterization of ELOVL1, a key enzyme in very long-chain fatty acid synthesis.
Enzymatic characterization of ELOVL1, a key enzyme in very long-chain fatty acid synthesis.
Biochimica et biophysica acta (2014-12-17)
Martin J A Schackmann, Rob Ofman, Inge M E Dijkstra, Ronald J A Wanders, Stephan Kemp
ABSTRACT
X-linked adrenoleukodystrophy (X-ALD) is a neurometabolic disease that is caused by mutations in the ABCD1 gene. ABCD1 protein deficiency impairs peroxisomal very long-chain fatty acid (VLCFA) degradation resulting in increased cytosolic VLCFA-CoA levels, which are further elongated by the VLCFA-specific elongase, ELOVL1. In adulthood, X-ALD most commonly manifests as a gradually progressive myelopathy (adrenomyeloneuropathy; AMN) without any curative or disease modifying treatments. We recently showed that bezafibrate reduces VLCFA accumulation in X-ALD fibroblasts by inhibiting ELOVL1. Although, in a clinical trial, bezafibrate was unable to lower VLCFA levels in plasma or lymphocytes in X-ALD patients, inhibition of ELOVL1 remains an attractive therapeutic option. In this study, we investigated the kinetic characteristics of ELOVL1 using X-ALD fibroblasts and microsomal fractions from ELOVL1 over-expressing HEK293 cell lines and analyzed the inhibition kinetics of a series of fibrates. Our data show that the CoA esters of bezafibrate and gemfibrozil reduce chain elongation by specifically inhibiting ELOVL1. These fibrates can therefore serve as lead compounds for the development of more potent and more specific inhibitors for ELOVL1.
Product Description
USP
Fenofibrate Related Compound B, United States Pharmacopeia (USP) Reference Standard
Supelco
Fenofibric acid, analytical standard
Supelco
Clofibric acid, analytical standard
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
2-(p-Chlorophenoxy)-2-methylpropionic acid, 97%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Behenic acid, analytical standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Behenic acid, 99%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis