Merck
  • Characterization of a broadly neutralizing monoclonal antibody that targets the fusion domain of group 2 influenza A virus hemagglutinin.
Characterization of a broadly neutralizing monoclonal antibody that targets the fusion domain of group 2 influenza A virus hemagglutinin.
Journal of virology (2014-09-12)
Gene S Tan, Peter S Lee, Ryan M B Hoffman, Beryl Mazel-Sanchez, Florian Krammer, Paul E Leon, Andrew B Ward, Ian A Wilson, Peter Palese
ABSTRACT
Due to continuous changes to its antigenic regions, influenza viruses can evade immune detection and cause a significant amount of morbidity and mortality around the world. Influenza vaccinations can protect against disease but must be annually reformulated to match the current circulating strains. In the development of a broad-spectrum influenza vaccine, the elucidation of conserved epitopes is paramount. To this end, we designed an immunization strategy in mice to boost the humoral response against conserved regions of the hemagglutinin (HA) glycoprotein. Of note, generation and identification of broadly neutralizing antibodies that target group 2 HAs are rare and thus far have yielded only a few monoclonal antibodies (MAbs). Here, we demonstrate that mouse MAb 9H10 has broad and potent in vitro neutralizing activity against H3 and H10 group 2 influenza A subtypes. In the mouse model, MAb 9H10 protects mice against two divergent mouse-adapted H3N2 strains, in both pre- and postexposure administration regimens. In vitro and cell-free assays suggest that MAb 9H10 inhibits viral replication by blocking HA-dependent fusion of the viral and endosomal membranes early in the replication cycle and by disrupting viral particle egress in the late stage of infection. Interestingly, electron microscopy reconstructions of MAb 9H10 bound to the HA reveal that it binds a similar binding footprint to MAbs CR8020 and CR8043. The influenza hemagglutinin is the major antigenic target of the humoral immune response. However, due to continuous antigenic changes that occur on the surface of this glycoprotein, influenza viruses can escape the immune system and cause significant disease to the host. Toward the development of broad-spectrum therapeutics and vaccines against influenza virus, elucidation of conserved regions of influenza viruses is crucial. Thus, defining these types of epitopes through the generation and characterization of broadly neutralizing monoclonal antibodies (MAbs) can greatly assist others in highlighting conserved regions of hemagglutinin. Here, we demonstrate that MAb 9H10 that targets the hemagglutinin stalk has broadly neutralizing activity against group 2 influenza A viruses in vitro and in vivo.
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 15 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 2 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 1 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 3 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 60 mg substrate per tablet
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 10 mg substrate per tablet
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%