Skip to Content
Merck
All Photos(2)

Key Documents

P7288

Sigma-Aldrich

o-Phenylenediamine dihydrochloride

chromogenic, tablet

Synonym(s):

1,2-Phenylenediamine tablet, OPD, OPD Tablet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3912045
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Pricing and availability is not currently available.

Product Name

o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet

Quality Level

form

tablet

mp

258 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Application

o-Phenylenediamine dihydrochloride has been used:
  • as a substrate for the eosinophil peroxidase (EPO) assay in bronchoalveolar lavage fluid (BALF)
  • in cell adhesion molecule-enzyme immunoassay (CAM-EIA) to analyze the amounts of E-selectin induced by test compounds (LPS or solutions and materials) on endothelial cells
  • in ultra-sensitive enzyme-linked immunosorbent assay (ELISA) to measure prolactin and luteinizing hormone (LH)

Reconstitution

Dissolve one tablet in 0.05 M phosphate-citrate buffer, pH 5.0, to the desired concentration (typically an OPD concentration of 0.4 mg/ml is used). Add 40 μl of fresh 30% hydrogen peroxide per 100 ml of substrate buffer solution, immediately prior to use. Tablets are individually packaged in foil packets.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Pero et al.
Molecular and cellular biochemistry, 193(1-2), 119-125 (1999-05-20)
Our laboratory has concentrated on the possible regulation the benzamides and nicotinamides may have on the processes of DNA repair and apoptosis. Recent reports have suggested that both apoptosis and inflammation are regulated by the transcription factor NF-kappaB. We have
D Sano et al.
Journal of applied microbiology, 109(6), 1923-1928 (2010-08-26)
Transmission routes of noroviruses, leading aetiological agents of acute gastroenteritis, are rarely verified when outbreaks occur. Because the destination of norovirus particles being firmly captured by micro-organisms could be totally different from that of those particles moving freely, micro-organisms with
Ronald E Unger et al.
Biomaterials, 35(10), 3180-3187 (2014-01-25)
Endotoxin, also known as lipopolysaccharide (LPS) produced by bacteria can be present in any liquid or on any biomaterial even if the material is sterile. Endotoxin in mammals can cause fever, inflammation, cell and tissue damage and irreversible septic shock
Hollian R Phillipps et al.
Journal of neuroendocrinology, 34(6), e13129-e13129 (2022-05-02)
Throughout the reproductive cycle in rodents, prolactin levels are generally low. In some species, including rats, a prolactin surge occurs on proestrus with peak concentrations coinciding with the preovulatory luteinizing hormone (LH) surge. In mice, however, there are conflicting reports
C A Ison
Methods in molecular medicine, 66, 317-329 (2001-01-01)
Neisseria meningitidis is an obligate human pathogen. When it interacts with the host, it can establish a commensal relationship or can, on a minority of occasions, invade and cause systemic disease. Protection against systemic disease, particularly for serogroup A and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service