Skip to Content
Merck
All Photos(2)

Key Documents

P6787

Sigma-Aldrich

o-Phenylenediamine dihydrochloride

chromogenic, tablet

Synonym(s):

1,2-Phenylenediamine tablet, OPD, OPD Tablet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3912045
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

o-Phenylenediamine dihydrochloride, tablet, 2 mg substrate per tablet

Quality Level

form

tablet

mp

258 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Reconstitution

Dissolve one tablet in 0.05 M phosphate-citrate buffer, pH 5.0, to the desired concentration (typically an OPD concentration of 0.4 mg/ml is used). Add 40 μl of fresh 30% hydrogen peroxide per 100 ml of substrate buffer solution, immediately prior to use. Tablets are individually packaged in foil packets.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah L Rowland et al.
The Journal of experimental medicine, 211(10), 1977-1991 (2014-09-03)
Plasmacytoid dendritic cells (pDCs) have long been implicated in the pathogenesis of lupus. However, this conclusion has been largely based on a correlative link between the copious production of IFN-α/β by pDCs and the IFN-α/β "signature" often seen in human
Florian Krammer et al.
Clinical and vaccine immunology : CVI, 21(8), 1153-1163 (2014-06-20)
Emerging H7N9 influenza virus infections in Asia have once more spurred the development of effective prepandemic H7 vaccines. However, many vaccines based on avian influenza viruses--including H7--are poorly immunogenic, as measured by traditional correlates of protection. Here we reevaluated sera
Cassandra Koole et al.
The Journal of pharmacology and experimental therapeutics, 353(1), 52-63 (2015-01-30)
The glucagon-like peptide-1 receptor (GLP-1R) is a class B G protein-coupled receptor that has a critical role in the regulation of glucose homeostasis, principally through the regulation of insulin secretion. The receptor system is highly complex, able to be activated
Tz-Chun Guo et al.
PloS one, 9(7), e100184-e100184 (2014-07-11)
Pancreas disease (PD) of Atlantic salmon is an emerging disease caused by Salmonid alphavirus (SAV) which mainly affects salmonid aquaculture in Western Europe. Although genome structure of SAV has been characterized and each individual viral protein has been identified, the
Gene S Tan et al.
Journal of virology, 88(23), 13580-13592 (2014-09-12)
Due to continuous changes to its antigenic regions, influenza viruses can evade immune detection and cause a significant amount of morbidity and mortality around the world. Influenza vaccinations can protect against disease but must be annually reformulated to match the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service