Merck
  • Non-linear increases in danazol exposure with dose in older vs. younger beagle dogs: the potential role of differences in bile salt concentration, thermodynamic activity, and formulation digestion.
Non-linear increases in danazol exposure with dose in older vs. younger beagle dogs: the potential role of differences in bile salt concentration, thermodynamic activity, and formulation digestion.
Pharmaceutical research (2014-01-31)
Mette U Anby, Hywel D Williams, Orlagh Feeney, Glenn A Edwards, Hassan Benameur, Colin W Pouton, Christopher J H Porter
ABSTRACT
To explore the possibility that age-related changes in physiology may result in differences in drug bioavailability after oral administration of lipid based formulations of danazol. Danazol absorption from lipid formulations with increasing drug load was examined in younger (9 months) and older (8 years) beagles. Age related changes to hepatic function were assessed via changes to systemic clearance and serum bile acid concentrations. Changes to lipolytic enzyme activity and intestinal bile salt concentration were evaluated using in vitro lipolysis. Drug exposure increased linearly with dose in younger animals. In older animals, bioavailability increased with increasing dose to a tipping point, beyond which bioavailability reduced (consistent with initiation of precipitation). No differences in hepatic function were apparent across cohorts. Changes to enzyme concentrations in lipolysis studies had little impact on drug precipitation/solubilisation. In contrast, higher bile salt concentrations better supported supersaturation at higher drug loads. Differences in animal cohort can have a significant impact on drug absorption from lipid based formulation. For danazol, bioavailability was enhanced under some circumstances in older animals. In vitro experiments suggest that this was unlikely to reflect changes to metabolism or lipolysis, but might be explained by increases in luminal bile salt/phospholipid concentrations in older animals.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Pricing and availability is not currently available.
Sigma-Aldrich
Progesterone, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Progesterone, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene sulfite, 98%
Pricing and availability is not currently available.
Supelco
Progesterone, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Pricing and availability is not currently available.
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Pricing and availability is not currently available.
Progesterone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene sulfite, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Pricing and availability is not currently available.
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Pricing and availability is not currently available.