Skip to Content
Merck
All Photos(1)

Documents

415413

Sigma-Aldrich

Sodium hydroxide solution

50% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316
PubChem Substance ID:
NACRES:
NA.21

Agency

suitable for DIN 38407-42
suitable for EPA 533
suitable for EPA 8327
suitable for EPA OTM-45
suitable for SM 4500 - NH3

Quality Level

vapor pressure

3 mmHg ( 37 °C)

form

liquid (viscous)

concentration

48-52% (by HCl, titration)
50% in H2O

technique(s)

HPLC: suitable

pH

14

density

1.515 g/mL at 25 °C

storage temp.

room temp

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide solution is a strong alkali. It is utilized in making cellophane, rayon, bleaches, dyes and drugs. It is also employed to abstract hydrogen sulfide from petroleum and to degrade lignin in wood in the paper industry.

Application

Sodium hydroxide solution may be used in the following processes:
  • As a dedoping reagent in the synthesis of graphene/polyaniline hybrid material.
  • As a solvent in the weight loss study of aluminum by corrosion.4
  • Alkaline reagent test for tannins & phenolic compounds.
  • Synthesis of single crystalline zinc stannate (Zn2SnO4 and ZnSnO3) nanoparticles.
  • Estimation of protein content and α-cellulose content in grain by-products.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single crystalline zinc stannate nanoparticles for efficient photo-electrochemical devices.
Miyauchi M, et al.
Chemical Communications (Cambridge, England), 46(9), 1529-1531 (2010)
Barley husk and coconut shell reinforced polypropylene composites: the effect of fibre physical, chemical and surface properties.
Bledzki AK, et al.
Composites Science and Technology, 70(5), 840-846 (2010)
Daly AO.
Sodium, 21-21 (2002)
A nanostructured graphene/polyaniline hybrid material for supercapacitors.
Wang H, et al.
Nanoscale, 2(10), 2164-2170 (2010)
Phytochemical investigation and chromatographic evaluation of the different extracts of tuber of Amorphaphallus paeoniifolius (Araceae).
De S and Ghosh AK.
International Journal of Pharmaceutical and Biomedical Research, 1(5), 150-157 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service