Merck
  • Behavioral and neuronal recording of the nucleus accumbens in adolescent rats following acute and repetitive exposure to methylphenidate.
Behavioral and neuronal recording of the nucleus accumbens in adolescent rats following acute and repetitive exposure to methylphenidate.
Journal of neurophysiology (2014-10-17)
Alexander Frolov, Cruz Reyes-Vasquez, Nachum Dafny
ABSTRACT
The nucleus accumbens (NAc) has been shown to play a key role in the brain's response to methylphenidate (MPD). The present study focuses on neuronal recording from this structure. The study postulates that repetitive exposure to the same dose of MPD will elicit in some rats behavioral sensitization and in others tolerance. Furthermore, the study postulates that NAc neuronal activity recorded from animals expressing behavioral tolerance after repetitive MPD exposure will be significantly different from NAc neuronal activity recorded from animals expressing behavioral sensitization after repetitive MPD exposure at doses of 0.6, 2.5, 5.0, and 10.0 mg/kg. To test this, behavioral and neuronal activity was recorded concomitantly from the NAc of freely behaving adolescent rats (postnatal day 40) before and after acute and repetitive administration of four different MPD doses. Comparing the acute MPD effect to the repetitive MPD effect revealed that the acute response to MPD exhibited dose-response characteristics: an increase in behavioral activity correlated with increasing MPD doses. On the other hand, following repetitive MPD exposure, some animals exhibited attenuated behavior (tolerance), while others exhibited further increases in the recorded behavior (sensitization). Moreover, the neuronal activity following repetitive MPD exposure recorded in animals exhibiting behavioral sensitization was significantly different from neuronal activity recorded in animals exhibiting behavioral tolerance. This implies that when studying the effects of repetitive MPD administration on adolescent rats, it is advisable to simultaneously record both neuronal and behavioral activity and to evaluate all data based on the animals' behavioral response to the repetitive MPD exposure.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Methylphenidate hydrochloride, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Methylphenidate hydrochloride
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)