Merck
  • Sharp-bounded zones link to the effect in planar chromatography-bioassay-mass spectrometry.
Sharp-bounded zones link to the effect in planar chromatography-bioassay-mass spectrometry.
Journal of chromatography. A (2014-08-26)
Ines Klingelhöfer, Gertrud E Morlock
ABSTRACT
The traditional direct bioautography workflow was substantially altered to yield narrow, sharp-bounded effective zones. For the first time, microorganisms quantitatively detected the single effective compounds in complex samples, separated in parallel on a planar chromatogram. This novel effect-directed workflow was demonstrated and optimized for the discovery of endocrine disrupting compounds (EDCs) reacting with the human estrogen receptor down to the femtogram-per-zone range, like 250fg/zone for 17β-estradiol (E2). For application volumes of up to 0.5mL, estrogen-effective compounds could directly be detected in complex samples at the ultratrace level (ng/kg-range). Sharp-bounded, estrogen-effective zones discovered were further characterized by direct elution into the mass spectrometer. HPTLC-ESI-MS mass spectra of (xeno)estrogens were shown for the first time. Owed to the substantially improved zone resolution, compound assignment was reliable and a comparison of the receptor affinities was conducted for six (xeno)estrogens. Also, long-term cell cultivation of the genetically modified yeast was demonstrated on the HPTLC plate. The optimized HPTLC-pYES workflow was proven for real food samples, exemplarily shown for beer. The general applicability of generating sharp-bounded zones was successfully proven by transfer of the fundamentally improved workflow to the Bacillus subtilis bioassay used for discovery of antibiotics in plant extracts. This new era of quantitative direct bioautography in combination with mass spectrometry will accelerate the scientific understanding in a wide application field via the streamlined access to fast and reliable information on effective components in complex samples.
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Estrone, ≥99%
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Ethinylestradiol, British Pharmacopoeia (BP) Assay Standard
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Estrone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Supelco
4-Nonylphenol, PESTANAL®, analytical standard
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%