Merck
  • Profiling of secondary metabolites in root exudates of Arabidopsis thaliana.
Profiling of secondary metabolites in root exudates of Arabidopsis thaliana.
Phytochemistry (2014-12-03)
Nadine Strehmel, Christoph Böttcher, Stephan Schmidt, Dierk Scheel
ABSTRACT
To explore the chemical composition of root exudates of the model plant Arabidopsis thaliana a workflow for nontargeted metabolite profiling of the semipolar fraction of root exudates was developed. It comprises hydroponic plant cultivation and sampling of root exudates under sterile conditions, sample preparation by solid-phase extraction and analysis by reversed-phase UPLC/ESI-QTOFMS. Following the established workflow, root exudates of six-week-old plants were profiled and a set of reproducibly occurring molecular features was compiled. To structurally elucidate the corresponding metabolites, accurate mass tandem mass spectrometry and on-line hydrogen/deuterium exchange were applied. Currently, a total of 103 compounds were detected and annotated by elemental composition of which more than 90 were structurally characterized or classified. Among them, 42 compounds were rigorously identified using an authenticated standard. The compounds identified so far include nucleosides, deoxynucleosides, aromatic amino acids, anabolites and catabolites of glucosinolates, dipeptides, indolics, salicylic and jasmonic acid catabolites, coumarins, mono-, di- and trilignols, hydroxycinnamic acid derivatives and oxylipins and exemplify the high chemical diversity of plant root exudates.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-methoxyindole-3-carboxaldehyde, AldrichCPR
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
15-Hydroxypentadecanoic acid, 97%
Supelco
2,4-D, PESTANAL®, analytical standard
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
2,4-D, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%