Skip to Content
Merck
All Photos(1)

Documents

49083

Supelco

2,4-D

PESTANAL®, analytical standard

Synonym(s):

2,4-Dichlorophenoxyacetic acid, 2,4-D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OCH2CO2H
CAS Number:
Molecular Weight:
221.04
Beilstein:
1214242
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor pressure

0.4 mmHg ( 160 °C)

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

vial of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

136-140 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)COc1ccc(Cl)cc1Cl

InChI

1S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

OVSKIKFHRZPJSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-D is a widely used, colorless and odorless herbicide, which can find applications in controlling broad-leaved weeds, woody plants in agriculture and also broad-leaves aquatic weeds. It can readily undergo degradation by microorganisms in soil and water.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A case-control study of non-Hodgkin's lymphoma and the herbicide 2, 4-dichlorophenoxyacetic acid (2, 4-D) in eastern Nebraska
Zahm HS, et al.
Epidemiology (Cambridge, Mass.), 349- 356 (1990)
Miller.HJ
Response of cotton to 2,4-D and related phenoxy herbicides, Volumes 1281-1292, 1281-1292 (1963)
Weiner.RE
Applications of Environmental Aquatic Chemistry: A Practical Guide, Second Edition, (2) (2008)
Manoharan Rajesh et al.
Protoplasma, 251(5), 1231-1243 (2014-03-19)
Podophyllum hexandrum Royle known as Indian mayapple is an important medicinal plant found only in higher altitudes (2,700 to 4,200 m) of the Himalayas. The highly valued anticancer drug Podophyllotoxin is obtained from the roots of this plant. Due to
Miguel Cacho Teixeira et al.
Trends in biotechnology, 25(8), 363-370 (2007-06-20)
Genomic information and tools are beginning to be used to increase our understanding of how organisms of all types interact with their environment. The study of the expression of all genes, at the genome, transcriptome, proteome and metabolome level, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service