Skip to Content
Merck
All Photos(1)

Key Documents

S-019

Supelco

Salicylic acid

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
774890
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

vapor density

4.8 (vs air)

vapor pressure

1 mmHg ( 114 °C)

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

GC/MS: suitable
gas chromatography (GC): suitable
liquid chromatography (LC): suitable

bp

211 °C (lit.)

mp

158-161 °C (lit.)

density

0.782 g/cm3 at 20 °C

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Salicylic acid, the major metabolite of aspirin, is also a phytohormone found in plants with roles in plant growth and development, photosynthesis, transpiration, ion uptake, and transport. This certified solution standard is suitable for numerous LC/MS and GC/MS applications including clinical and diagnostic testing, pharmaceutical research and phytochemical testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shifeng Cao et al.
Food chemistry, 134(4), 1715-1718 (2013-02-28)
The effect of elicitors associated with host defence on betacyanin accumulation in Amaranthus mangostanus seedlings was investigated. Under the conditions of the experiments, betacyanin accumulation was generally enhanced by light. Methyl jasmonate (MeJA) treatment increased betacyanin synthesis in a concentration-dependent
E Benfeldt et al.
Acta dermato-venereologica, 79(5), 338-342 (1999-09-24)
Our aim was to simultaneously investigate 2 techniques for in vivo sampling of peripheral compartment pharmacokinetics after systemic administration of acetylsalicylic acid. Ten volunteers were given 2 g acetylsalicylic acid orally. Blood samples and dialysates from 4 microdialysis probes inserted
Tsung-Meng Wu et al.
Plant molecular biology, 83(4-5), 379-390 (2013-06-21)
Glutathione reductases (GRs) are important components of the antioxidant machinery that plants use to respond against abiotic stresses. In rice, one cytosolic and two chloroplastic GR isoforms have been identified. In this work, we describe the cloning and characterization of
Dorothea Ellinger et al.
Plant physiology, 161(3), 1433-1444 (2013-01-22)
A common response by plants to fungal attack is deposition of callose, a (1,3)-β-glucan polymer, in the form of cell wall thickenings called papillae, at site of wall penetration. While it has been generally believed that the papillae provide a
Yunlong Du et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7946-7951 (2013-04-25)
Removal of cargos from the cell surface via endocytosis is an efficient mechanism to regulate activities of plasma membrane (PM)-resident proteins, such as receptors or transporters. Salicylic acid (SA) is an important plant hormone that is traditionally associated with pathogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service