Merck
  • A semi-probabilistic modelling approach for the estimation of dietary exposure to phthalates in the Belgian adult population.
A semi-probabilistic modelling approach for the estimation of dietary exposure to phthalates in the Belgian adult population.
Environment international (2014-08-13)
T Fierens, A Standaert, C Cornelis, I Sioen, S De Henauw, H Willems, M Bellemans, M De Maeyer, M Van Holderbeke
ABSTRACT
In this study, a semi-probabilistic modelling approach was applied for the estimation of the long-term human dietary exposure to phthalates--one of world's most used families of plasticisers. Four phthalate compounds were considered: diethyl phthalate (DEP), di-n-butyl phthalate (DnBP), benzylbutyl phthalate (BBP) and di(2-ethylhexyl) phthalate (DEHP). Intake estimates were calculated for the Belgian adult population and several subgroups of this population for two considered scenarios using an extended version of the EN-forc model. The highest intake rates were found for DEHP, followed by DnBP, BBP and DEP. In the Belgian adult population, men and young adults generally had the highest dietary phthalate intake estimates. Nevertheless, predicted dietary intake rates for all four investigated phthalates were far below the corresponding tolerable daily intake (TDI) values (i.e. P99 intake values were 6.4% of the TDI at most), which is reassuring because adults are also exposed to phthalates via other contamination pathways (e.g. dust ingestion and inhalation). The food groups contributing most to the dietary exposure were grains and grain-based products for DEP, milk and dairy products for DnBP, meat and meat products or grains and grain-based products (depending on the scenario) for BBP and meat and meat products for DEHP. Comparison of the predicted intake results based on modelled phthalate concentrations in food products with intake estimates from other surveys (mostly based on measured concentrations) showed that the extended version of the EN-forc model is a suitable semi-probabilistic tool for the estimation and evaluation of the long-term dietary intake of phthalates in humans.
Product Description
Supelco
Di-n-butyl phthalate solution, certified reference material, 5000 μg/mL in methanol
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore, for ion-selective electrodes
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Diethyl phthalate, ≥99%
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Diethyl phthalate, 99.5%
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
Diethyl phthalate, European Pharmacopoeia (EP) Reference Standard