Skip to Content
Merck
All Photos(1)

Key Documents

W512206

Sigma-Aldrich

Diethyl phthalate

≥99%

Synonym(s):

Anozol, Diethyl benzene-1,2-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1912500
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
odorless
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

Assay

≥99%

form

liquid

autoignition temp.

854 °F

expl. lim.

0.75 %, 187 °F

refractive index

n20/D 1.502 (lit.)

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diethyl phthalate is commonly used as a vehicle for fragrances and cosmetic ingredients.[1] It is one of the volatile compounds found in rice cultivars[2] and starfruit.[3]

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile profiles of aromatic and non-aromatic rice cultivars using SPME/GC-MS
Bryant RJ and McClung AM.
Food Chemistry, 124(2), 501-513 (2011)
Volatile components of starfruit
MacLeod G and Ames JM
Phytochemistry, 29(1), 165-172 (1990)
Raphael J Witorsch et al.
Critical reviews in toxicology, 40 Suppl 3, 1-30 (2010-10-12)
This article reviews laboratory and epidemiological research into the endocrine disruptive effects of components of personal care products, namely, phthalate esters, parabens, ultraviolet (UV) filters, polycyclic musks, and antimicrobials. High doses of phthalates in utero can produce “phthalate syndrome,” demasculinizing
Kashappa-Goud H Desai et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(1), 62-74 (2012-10-30)
Herein, we describe the detailed development of a simple and effective method to microencapsulate vaccine antigens in poly(lactic-co-glycolic acid) (PLGA) by simple mixing of preformed active self-microencapsulating (SM) PLGA microspheres in a low concentration aqueous antigen solution at modest temperature
Lee-Ju Cheng et al.
Aquatic toxicology (Amsterdam, Netherlands), 109, 166-175 (2011-10-28)
The toxicity and effects of diethyl phthalate (DEP), a potent allelochemical, on the growth of greater duckweed were studied. Biochemical analyses and physiological methods were combined to investigate oxidative stress, adverse effects and their mechanisms in greater duckweeds grown in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service