Skip to Content
Merck
All Photos(1)

Key Documents

40066

Supelco

Di-n-butyl phthalate solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

O(CCCC)C(=O)c1c(cccc1)C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3

InChI key

DOIRQSBPFJWKBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di-n-butyl phthalates (DnBPs) are mainly used in personal-care products like perfumes, aftershaves, shampoos, make up and nail products, paints, etc.

Application

DnBP has been used as a certified reference material (CRM) for the determination of the analyte in cleaning and personal care products by liquid chromatography-mass spectrometry (LC-MS) and as a reference standard for the identification of the analyte in tea beverages by ionic liquid-based hollow fiber liquid phase microextraction (IL-HF-PLME), coupled with high-performance liquid chromatography-diode array detection (HPLC-DAD). It may be used as a reference standard for the analysis of the analyte in food samples and as urinary metabolites in pregnant women by LC-MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Repr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method development for the analysis of phthalate esters in tea beverages by ionic liquid hollow fibre liquid-phase microextraction and liquid chromatographic detection
Wang J, et al.
Food Control, 67, 278-284 (2016)
Determination of 12 urinary phthalate metabolites in Norwegian pregnant women by core-shell high performance liquid chromatography with on-line solid-phase extraction, column switching and tandem mass spectrometry
Sabaredzovic A, et al.
Journal of Chromatography. B, Biomedical Applications, 1002, 343-352 (2015)
Determination of 23 phthalic acid esters in food by liquid chromatography tandem mass spectrometry
Xu D, et al.
Journal of Chromatography A, 1324, 49-56 (2014)
Carl Simonsson et al.
Toxicology and applied pharmacology, 264(1), 114-120 (2012-07-31)
Allergic contact dermatitis (ACD) is caused by low-molecular weight compounds called haptens. It has been shown that the potency of haptens can depend on the formulation in which they are applied on the skin. Specifically the sensitization potency of isothiocyanates
Susan Y Euling et al.
Toxicology and applied pharmacology, 271(3), 349-362 (2011-07-13)
An evaluation of the toxicogenomic data set for dibutyl phthalate (DBP) and male reproductive developmental effects was performed as part of a larger case study to test an approach for incorporating genomic data in risk assessment. The DBP toxicogenomic data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service