Merck
  • Ascorbic acid: a review of its chemistry and reactivity in relation to a wine environment.
Ascorbic acid: a review of its chemistry and reactivity in relation to a wine environment.
Critical reviews in food science and nutrition (2011-09-21)
Marc P Bradshaw, Celia Barril, Andrew C Clark, Paul D Prenzler, Geoffrey R Scollary
ABSTRACT
Extensive reviews of research are available on the use of ascorbic acid, and its consequent degradation pathways, in physiological conditions or food matrices. However, very little information can be found for wine-related systems. This review highlights the relevant chemistry and reactivity of ascorbic acid with a focus on its behavior and potential behavior in a wine environment. The review describes the use of ascorbic acid as a complementary antioxidant preservative to sulfur dioxide along with the metal-catalyzed and radical-dependent manner by which it achieves this role. The relevant degradation products of ascorbic acid in aerobic and anaerobic conditions are presented as well as the interaction of these degradation products with sulfur dioxide and other wine-relevant sulfur compounds. Limitations in existing knowledge, especially regarding the crossover between the antioxidant and pro-oxidant roles of ascorbic acid, are identified.
Product Description
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard