Merck
  • Consequences of biodiversity loss for litter decomposition across biomes.
Consequences of biodiversity loss for litter decomposition across biomes.
Nature (2014-05-09)
I Tanya Handa, Rien Aerts, Frank Berendse, Matty P Berg, Andreas Bruder, Olaf Butenschoen, Eric Chauvet, Mark O Gessner, Jérémy Jabiol, Marika Makkonen, Brendan G McKie, Björn Malmqvist, Edwin T H M Peeters, Stefan Scheu, Bernhard Schmid, Jasper van Ruijven, Veronique C A Vos, Stephan Hättenschwiler
ABSTRACT
The decomposition of dead organic matter is a major determinant of carbon and nutrient cycling in ecosystems, and of carbon fluxes between the biosphere and the atmosphere. Decomposition is driven by a vast diversity of organisms that are structured in complex food webs. Identifying the mechanisms underlying the effects of biodiversity on decomposition is critical given the rapid loss of species worldwide and the effects of this loss on human well-being. Yet despite comprehensive syntheses of studies on how biodiversity affects litter decomposition, key questions remain, including when, where and how biodiversity has a role and whether general patterns and mechanisms occur across ecosystems and different functional types of organism. Here, in field experiments across five terrestrial and aquatic locations, ranging from the subarctic to the tropics, we show that reducing the functional diversity of decomposer organisms and plant litter types slowed the cycling of litter carbon and nitrogen. Moreover, we found evidence of nitrogen transfer from the litter of nitrogen-fixing plants to that of rapidly decomposing plants, but not between other plant functional types, highlighting that specific interactions in litter mixtures control carbon and nitrogen cycling during decomposition. The emergence of this general mechanism and the coherence of patterns across contrasting terrestrial and aquatic ecosystems suggest that biodiversity loss has consistent consequences for litter decomposition and the cycling of major elements on broad spatial scales.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Supelco
Activated charcoal, powder
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 1.5mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.050μm, specific density 10μg/cm2, temporary glass support, 99.997%
Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon, rod, 300mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, rod, 50mm, diameter 25mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 100mm, diameter 2.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 13.0mm, graphite, 100%