Merck
  • Flagella interacting with a carbon nanowire with the variation of time and initial temperature.
Flagella interacting with a carbon nanowire with the variation of time and initial temperature.
Journal of nanoscience and nanotechnology (2014-04-18)
Elson C Santos, Alessandro J G Santos, Abel F G Neto, Antonio M J C Neto, Rosivaldo S Borges, Teodorico C Ramalho
ABSTRACT
The system proposed consists of a flagellum relaxing around a static carbon nanowire to mimics behavior of a natural flagellum moving with damped harmonic motion along a wire under van der Waals and electrostatic forces. This flagellum is composed of a C20 nanosphere with different sizes of his tail formed by hydrocarbons. The thermodynamic properties such as molar entropy variation, as well as molar heat dissipation, efficiency and speed were obtained to evaluate which system is most stable by using the variable temperature. This system has a number of carbon atoms ranging from 103-110, with a maximum of 300 ps for each simulation. We had simulated molar entropy variation, energies and efficiency changing with time and initial temperature. The results indicate that among the systems studied, the flagellum with five carbon atoms achieved greater stability and better results in this search.
Product Description
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Supelco
Activated charcoal, powder
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated charcoal, puriss. p.a., powder
Millipore
Activated charcoal, suitable for GC
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Carbon, foil, 150x150mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, annealed, 99.997%
Carbon, foil, 12mm disks, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, not light tested, 150x150mm, thickness 0.125mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 0.2mm, flexible graphite, 99.8%
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Carbon, foil, 100x100mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, tube, graphite, 150mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon