Skip to Content
Merck
All Photos(2)

Key Documents

GF46470944

Carbon

foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%

Synonym(s):

Activated charcoal, C 000315, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.8%

form

foil

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 464-709-44

resistivity

1375 μΩ-cm, 20°C (graphite)

size × thickness

100 × 100 mm × 0.35 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doubly differential secondary-electron yields following 8-MeV/u U68+- and 3.5-MeV/u U38+-ion impact on a thin carbon-foil target.
Schneider et al.
Physical review. A, Atomic, molecular, and optical physics, 47(5), 3945-3950 (1993-05-01)
I Dietrich et al.
Ultramicroscopy, 3(2), 185-189 (1978-01-01)
Radiation damage on a holey carbon foil was investigated in an electron microscope with a superconducting lens system, where the temperature of the specimen and its environment initially was 4 K. Due to an electron dose of 2 X 10(4)
Investigation of delta -electron emission in collisions of highly charged fast Ne projectiles with carbon-foil targets.
Schiwietz et al.
Physical review. B, Condensed matter, 41(10), 6262-6271 (1990-04-01)
M C Ledbetter et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(1), 85-88 (1987-01-01)
Cluster ions containing 50, 100, or 200 water molecules and one proton are accelerated to over 275 kV and impacted on thin carbon films on which tobacco mosaic virus has been dispersed. After bombardment the films are examined with a
Z oscillation of mean charges of energetic ions emerging from a carbon foil: Correlation with the shell structure of ions.
Shima et al.
Physical review. A, General physics, 39(8), 4316-4318 (1989-04-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service